Learn More
CNOT1 Mouse anti-Human, CoraLite 488, Clone: 1A10A11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66507-100UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CNOT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CNOT1. This antibody recognizes human antigen. The CNOT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifica
| CNOT1 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Immunofluorescence | |
| 1A10A11 | |
| CoraLite 488 | |
| 100 μL | |
| 23019 | |
| IgG1 |