Learn More
CNOT9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 22503-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a member of the highly conserved RCD1 protein family. The encoded protein is a transcriptional cofactor and a core protein of the CCR4-NOT complex. It may be involved in signal transduction as well as retinoic acid-regulated cell differentiation and development. Alternatively spliced transcript variants have been described for this gene.Specifica
| CNOT9 | |
| Polyclonal | |
| Unconjugated | |
| CNOT9 | |
| CNOT9, Rcd 1, RCD1, RCD1+, RQCD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 301513, 58184, 9125 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5PQL2, Q92600, Q9JKY0 | |
| Cnot9 | |
| CNOT9 Fusion Protein Ag18197 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |