Learn More
Aalborg™ CO2 Direct Flow Tube
Codice prodotto. 11862250
Click to view available options
Velocità di flusso dell'aria:
1 L/min.
100 mL/min.
300 mL/min.
6 L/min.
Larghezza (sistema metrico):
65 mm
150 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 11862250
Marca: Aalborg™ FT05214GLVN
Aalborg™ CO2 Direct Flow Tube completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifica
Tipo CO2 Direct Flow Tube
Larghezza (sistema metrico) 65 mm
Velocità di flusso dell'aria 1 L/min.