Learn More
TFA Dostmann CO2 Monitor AIRCO2NTROL COACH
Codice prodotto. 17941662
Cambia vista
Click to view available options
Analiti rilevabili:
Carbon Dioxide
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Analiti rilevabili Dimensione della confezione
17941662 Carbon Dioxide Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17941662 Fornitore TFA Dostmann N. del fornitore 31.5009.02
  • ONLINE EXCLUSIVE PRICE
    105.65€
    85.22€ / pezzi 1
    Risparmia 20.43€
    19% Sconto
Spedizione stimata: 23-11-2026
The CO2 Monitor AIRCO2NTROL COACH shows the current CO2 content of the room air, the temperature, humidity, and the time. Depending on the settings, it displays the current CO2 readings through the traffic light colours. The graph clearly shows the CO2 levels development of the last 24 hours.
Who breathes good air is healthier, happier and can do more. Headaches, dizziness and permanent fatigue are often due to a high concentration of CO2 in the air. TFA's AIRCO2NTROL COACH CO2 monitor helps you monitor how stale and contaminated the air is in indoor spaces where many people are present such as in schools, offices and public buildings. Timely and energy-efficient ventilation ensures a healthy indoor climate. Furthermore, a high CO2 value is considered an indicator for a possibly virus-contaminated aerosol concentration in the room air. The more exhaled air there is in the room, the higher the probability of infection via aerosols. The monitor shows on the display the current CO2 content of the room air, the temperature, humidity and the time.
The display's illumination color can be used to quickly evaluate and interpret the current CO2 readings. Depending on the setting, the illumination directly signals the current air quality in the traffic light colors (green, orange, red). Alternatively, the red display illumination can be used as an optical signal in the event of an alarm when a certain limit has been exceeded. As a further variant, you can choose a solid color from seven shades or switch off the illumination completely.
The three smiley icons serve as a comfort-level zone indicator for the measured values of the last 24 hours. If the reading is at the level of the happy smiley, the air quality is ideal to good. In addition, the graph clearly shows the CO2 levels development of the last 24 hours. The power is supplied via a suitable USB power source (PC, laptop).
  • High-quality NDIR Dual Beam Sensor
  • CO2 level measurement with 24-hours graph
  • Micro-USB cable inclusive
  • For the monitoring of CO2 concentration in buildings, where people are present, e.g. schools, offices, public buildings
  • Backlight color shows CO2 level, alarm or preferred color
  • Indication of time
  • Indoor temperature and humidity
Specifica
Analiti rilevabili Carbon Dioxide
Tensione 5 V