missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor III/Tissue Factor Antibody (323519), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB23391MFV500
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Coagulation Factor III/Tissue Factor Monoclonal antibody specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifica
| Coagulation Factor III/Tissue Factor | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 2152 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, CyTOF | |
| 323519 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III, Gly34-Glu251, Accession # P13726 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto