missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VII Antibody (LH7.2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-08278AF405
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Collagen VII Monoclonal antibody specifically detects Collagen VII in Human, Bovine, Equine, Guinea Pig, Goat, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifica
| Collagen VII | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Collagen 7, collagen alpha-1(VII) chain, collagen VII, alpha-1 polypeptide, collagen, type VII, alpha 1, Collagen-7, EBD1, EBDCT, EBR1, epidermolysis bullosa, dystrophic, dominant and recessive, LC collagen, long chain collagen, Long-chain collagen | |
| Recombinant full-length human Collagen VII protein (Uniprot: Q02388) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Extracellular Matrix | |
| 1294 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| LH7.2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Equine, Guinea Pig, Goat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto