Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel with Removable Glass Jacket
missing translation for 'orderingAttributeHoverText'
Lunghezza (sistema metrico):
200 mm
300 mm
500 mm
800 mm
missing translation for 'unitSize'
Pezzo
Description
- Columns acc. to Hempel
- With removable glass jacket
- For packings like Raschig rings or glass beads
- With NS socket and NS cone
- With removable glass jacket
- Made of DURAN™ tubing
Specifications
Specifications
| Include | Removable glass jacket |
| Lunghezza (sistema metrico) | 200 mm |
| Giunto rastremato standard | NS 14/23 |