Learn More
Abeba™ Comfort 36872 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB A, E SRB
- Velours
- Black
- Slip resistance PU outsole sole
- Replaceable Reflexor insole with nubs (article no. 3572)
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Instep area with elastic band
- Buckle fastening
- Hand-sewn seam
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, E SRB |
| Colore | Black |
| Materiale rivestimento interno | Microfiber |
| Materiale | Velour |
| Size | 41 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 36872 |