missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Antibody (135727), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals FAB7841X
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifica
| Common gamma Chain/IL-2 R gamma | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 135727 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Common gamma Chain/IL-2 R gamma , Met1-Ala263, Accession # P34902 | |
| 0.5 mL | |
| Cancer | |
| 3561 | |
| Store at 4C in the dark. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto