Learn More
Nabertherm™ Compact Tube Furnace with B510 Controller
Codice prodotto. 17328089 Sfoglia Tutto Nabertherm Prodotti
Click to view available options
Diametro (sistema metrico) camera d'aria:
100 mm
120 mm
170 mm
Zona riscaldata:
500 mm
750 mm
1000 mm
Temperatura (sistema metrico):
1200°C
1300°C
Dimensione della confezione:
Pezzo
Codice prodotto. 17328089
Marca: Nabertherm™ R322L3AN
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Single-zoned design as standard with controller B 510
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifica
Tipo di unità di controllo B510
Zona riscaldata 500 mm
Temperatura (sistema metrico) 1300°C
Tipo Compact Tube Furnace
Diametro (sistema metrico) camera d'aria 120 mm