missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3 Antibody (11H9) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 11 publications
Marca: Novus Biologicals NB200-540SS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Complement C3 Monoclonal specifically detects Complement C3 in Mouse, E. coli samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay, Flow (Intracellular), CyTOF-ready.
Specifica
| Complement C3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1-2 ug/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay 0.5 ug/well in PBS, | |
| P01027 | |
| C3 | |
| This Complement C3 Antibody (11H9) was developed against C57BL/6 thymocytes saturated with rat anti-Thy-1 monoclonal antibody of IgG2b subclass (RmT1). | |
| 0.025 mL | |
| Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction | |
| 718 | |
| Mouse, E. coli | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 11H9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse Complement C3 and its activation products, C3b, iC3b, C3d and C3dg | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto