Learn More
Invitrogen™ Complement C3 Monoclonal Antibody (11H9)
GREENER_CHOICE
Rat Monoclonal Antibody
Marca:  Invitrogen™ MA140046
Codice prodotto. 11519672
  • 920.00€ / 100µg
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
MA1-40046 detects C3 from mouse samples. MA1-40046 has been successfully used in flow cytometry, immunohistochemistry (frozen, paraffin), ELISA, immunofluorescence, immunoprecipitation, and Western blot applications. ELISA: Antibody was used as coating 0.5 μg/well in PBS. The MA1-40046 immunogen is mouse C3 acute phase protein.
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Complement C3
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
P01027
C3
C57BL/6 thymocytes saturated with rat anti-Thy-1 monoclonal antibody of IgG2b subclass (RmT1).
100 μg
Primary
Mouse
Antibody
IgG2a
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
11H9
Unconjugated
C3
Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; I79_008227; LOC100060539; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562)
Rat
Protein G
RUO
12266
4°C