missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C3c Monoclonal Antibody (4)

Rat Monoclonal Antibody
Marca: Invitrogen™ MA533336
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody recognizes a conformational epitope in C3c, C3b and iC3b. Product is supplied at 1 mL (100 μL/mL).
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
Specifica
| Complement C3c | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| P01024 | |
| C3 | |
| Protein G | |
| RUO | |
| 718 | |
| 4°C | |
| Liquid |
| Immunoprecipitation | |
| 4 | |
| Unconjugated | |
| C3 | |
| Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component 3d; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; LOC101108131; LOW QUALITY PROTEIN: complement C3; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562) | |
| Rat | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto