Learn More
Complement C4c Antibody (99-72-18), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-12091MFV610
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Complement C4c Monoclonal antibody specifically detects Complement C4c in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifica
| Complement C4c | |
| RUO | |
| Primary |
| ELISA | |
| Immunology, Innate Immunity |