missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H-related 5/CFHR5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals H00081494-D01P
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Complement Factor H-related 5/CFHR5 Polyclonal antibody specifically detects Complement Factor H-related 5/CFHR5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifica
| Complement Factor H-related 5/CFHR5 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| CFHL5factor H-related protein 5, complement factor H-related 5, FHR-5complement factor H-related protein 5, FHR5MGC133240, FLJ10549 | |
| CFHR5 (NP_110414.1, 1 a.a. - 569 a.a.) full-length human protein. MLLLFSVILISWVSTVGGEGTLCDFPKIHHGFLYDEEDYNPFSQVPTGEVFYYSCEYNFVSPSKSFWTRITCTEEGWSPTPKCLRMCSFPFVKNGHSESSGLIHLEGDTVQIICNTGYSLQNNEKNISCVERGWSTPPICSFTKGECHVPILEANVDAQPKKESYKVGDVLKFSCRKNLIRVGSDSVQCYQFGWSPNFPTCKGQVRSCGPPPQLSNGEVKEIRKEEYGHNEVVEYDCNPNFIINGPKKIQCVDGEWTTLPTCVEQVKTCGYIPELEYGYVQPSVPPYQHGVSVEVNCRNEYAMIGNNMITCINGIWTELPMCVATHQLKRCKIAGVNIKTLLKLSGKEFNHNSRIRYRCSDIFRYRHSVCINGKWNPEVDCTEKREQFCPPPPQIPNAQNMTTTVNYQDGEKVAVLCKENYLLPEAKEIVCKDGRWQSLPRCVESTAYCGPPPSINNGDTTSFPLSVYPPGSTVTYRCQSFYKLQGSVTVTCRNKQWSEPPRCLDPCVVSEENMNKNNIQLKWRNDGKLYAKTGDAVEFQCKFPHKAMISSPPFRAICQEGKFEYPICE | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| NP_110414.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 81494 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto