Learn More
Quickfit™ Single Cone with Long Tip
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
34/35
40/38
43027
45/40
55/44
7/16
Spessore (sistema metrico) parete:
1mm
1.5mm
1.8mm
2mm
2.3mm
2.5mm
Dimensione della confezione:
pezzi 10
pezzi 50
Descrizione
- Useful for construction of stirrer guides
- Unbadged and free from ceramic markings
- Avoids problem of badge charring at high temperatures which can give an untidy appearance
- Long tip cones manufactured with minimum shank length of 90mm
Specifica
Specifica
| Quantità | 10Pack |
| Tipo | Single Cone with Long Tip |
| Cone Size | 19/26 |
| Spessore (sistema metrico) parete | 1.8mm |