Learn More
Lenz Laborglasintrumente Cones with Drip Tip
Codice prodotto. 10418982
Click to view available options
Joint Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
Dimensione della confezione:
pezzi 10
Codice prodotto. 10418982
Marca: Lenz Laborglasintrumente 01100429
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12249 form F
  • Conical ground cones
  • Made of DURAN™ tubing
Specifica
Joint Size NS 29/32
Spessore (sistema metrico) parete 2 mm
Diameter (Metric) Outer Tube 26 mm
Materiale Borosilicate Glass (3.3)