Learn More
Lenz Laborglasintrumente Cones with Tip
Codice prodotto. 10234291
Click to view available options
Joint Size:
NS 10/19
NS 12/21
NS 34/35
NS 40/38
NS 50/42
NS 55/44
NS 7/16
NS 71/51
NS 85/55
Dimensione della confezione:
Pezzo
pezzi 10
Codice prodotto. 10234291
Marca: Lenz Laborglasintrumente 01100355
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12249 form G and in addition to standard
  • Conical ground cones
  • Made of DURAN™ tubing
Specifica
Quantità 1 Ea.
Joint Size NS 55/44
Spessore (sistema metrico) parete 2.5 mm
Diameter (Metric) Outer Tube 50 mm
Materiale Borosilicate Glass (3.3)