Learn More
Lenz Laborglasintrumente Cones with Tip
Codice prodotto. 10375731
Click to view available options
Joint Size:
NS 10/19
NS 12/21
NS 34/35
NS 40/38
NS 50/42
NS 55/44
NS 7/16
NS 71/51
NS 85/55
Dimensione della confezione:
Pezzo
pezzi 10
Codice prodotto. 10375731
Marca: Lenz Laborglasintrumente 01100350
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12249 form G and in addition to standard
  • Conical ground cones
  • Made of DURAN™ tubing
Specifica
Quantità 1 Ea.
Joint Size NS 50/42
Spessore (sistema metrico) parete 2.3 mm
Diameter (Metric) Outer Tube 46 mm
Materiale Borosilicate Glass (3.3)