Learn More
Lenz Laborglasintrumente Cones with Tip
Codice prodotto. 10532681
Click to view available options
Joint Size:
NS 10/19
NS 12/21
NS 34/35
NS 40/38
NS 50/42
NS 55/44
NS 7/16
NS 71/51
NS 85/55
Dimensione della confezione:
Pezzo
pezzi 10
Codice prodotto. 10532681
Marca: Lenz Laborglasintrumente 01100334
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN 12249 form G and in addition to standard
  • Conical ground cones
  • Made of DURAN™ tubing
Specifica
Quantità 10/Pk.
Joint Size NS 34/35
Spessore (sistema metrico) parete 2 mm
Diameter (Metric) Outer Tube 30 mm
Materiale Borosilicate Glass (3.3)