Learn More
Thermo Scientific Chemicals Al-23 Crucible, Conical;Ht (mm), 45;Top OD (mm), 35;Top ID (mm), 31;Base OD (mm), 18;Volume (ml), 23
Click to view available options
Capacità (sistema metrico):
4 mL
5 mL
8 mL
9 mL
23 mL
40 mL
49 mL
75 mL
125 mL
160 mL
Diametro (sistema metrico) esterno:
17mm
21mm
22mm
26mm
31mm
40mm
45mm
53mm
67mm
80mm
Diametro (sistema metrico) interno:
20mm
24mm
25mm
30mm
40mm
45mm
55mm
60mm
65mm
90mm
Altezza (sistema metrico):
20mm (Top), 11mm (Base)
25mm (Top), 13mm (Base)
30mm (Top), 15mm (Base)
35mm (Top), 18mm (Base)
45mm (Top), 22mm (Base)
50mm (Top), 25mm (Base)
60mm (Top), 30mm (Base)
73mm (Top), 35mm (Base)
88mm (Top), 44mm (Base)
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Diametro (sistema metrico) interno | 25mm |
| Materiale | Al-23 |
| Quantità | 1 Piece |
| Capacità (sistema metrico) | 4 mL |
| Diametro (sistema metrico) esterno | 17mm |
| Altezza (sistema metrico) | 20mm (Top), 11mm (Base) |
| Product Type | Conical Al-23 Crucibles |