Learn More
BOLA Connection Bolts
Click to view available options
Include:
Fingertight Nut, Single Ferrule
Hex-head Nut, Double Ferrule
Dimensione della confezione:
Pezzo
Descrizione
- Connection bolt and sealing cone made of PEEK, suitable for female thread UNF 10-32 G.
- Exchangeable sealing cone available separately, knurled nut reusable.
- Suitable for pressures up to max.
- 280 bar
Specifica
Specifica
| Diametro (sistema imperiale) esterno | 1.6 in. |
| Include | Hex-head Nut, Double Ferrule |
| Autoclavabile | Autoclavable |