Learn More
Gilson™ Connection Kit
Sfoglia Tutto Gilson Prodotti
Click to view available options
Diameter (Metric) Outside:
3 mm
4 mm
Dimensione della confezione:
pezzi 5
Descrizione
- Connector kit accessories for the MINIPULS™ 3 and Evolution peristaltic pumps come in a variety of types: metal sleeve connectors for small bore tubing, connector kits for flanged tubing, or plastic connectors for large bore tubing.
- These kits contain five connectors each.
- The connectors are available in a three sizes to fit a variety of tubing of different inner diameters.
Specifica
Specifica
| Diameter (Metric) Outside | 4 mm |
| Diameter (Metric) Inside | 2.0 to 2.3 mm |
| Quantità | 5 Set |
| Tipo | Connection Kit |