Learn More
Thermo Scientific™ Consumables Kits for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Codice prodotto. p-10421570 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
iCAP Q/Qnova ICP-MS Systems
Dimensione della confezione:
Pezzo
Codice prodotto. 17868504
Marca: Thermo Scientific™ 1324980
Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Consumables Kits include consumables for continual regular maintenance, to ensure your system is achieving maximum performance.
Use the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Consumables Kits, including consumables for continual regular maintenance, to ensure your system is achieving maximum performance.
Includes the following consumables to run the ICP-MS system for one year:
  • Quartz torch
  • Quartz injector, 5 mm
  • Replacement sample and skimmer cones (incl. spare sample cone gaskets)
  • For iCAP Qnova: Replacement inserts (2.8 mm, 3.5 mm, 4.5 mm)
  • Tubing for sample introduction (includes peristaltic pump tubing)
Specifica
Tipo Consumables Kit
Descrizione iCAP Q/Qnova Series Consumables Kit
Da utilizzare con (apparecchiatura) iCAP Q/Qnova ICP-MS Systems
Include Torch, Inserts, Cones and Sample Introduction Tubing