Learn More
Corning™ Moduli piastra e serbatoio CoolRack™
BIOPHARMA
Codice prodotto. 15552841
Click to view available options
Capacità:
1 Flat Bottom Plate, 1 SBS-compatible
1 U-bottom Microplate, SBS compatible
Reagent Reservoir (55 mL)
Dimensione della confezione:
Pezzo
Codice prodotto. 15552841
Marca: Corning™ 432070
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Garanzia di uniformità di temperatura (±0.1 °C) per tutte le provette per raffreddamento, surgelamento, riscaldamento o scongelamento.
Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.
  • CoolSink modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
  • All CoolSink modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • All modules are compatible with all temperature sources.
  • SBS-compatible
TRUSTED_SUSTAINABILITY
Specifica
Capacità 1 Flat Bottom Plate, 1 SBS-compatible
Progettazione CoolSink XT 96F
Tipo CoolSink Plate and Reservoir Module