Learn More
Corning™ CoolRack™ V Blood Collection Tube Modules
Click to view available options
Capacità:
12 (13 or 16 mm Tubes)
9 (13 x 100 mm Blood Tubes or 5 mL Cryogenic Vials)
9 (13 x 75 mm Blood Tubes)
9 (16 x 100 mm Blood Tubes)
Progettazione:
CoolRack V
CoolRack V13
CoolRack V16
CoolRack VS13
Dimensione della confezione:
Pezzo
Descrizione
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to > 100°C and it will rapidly adapt to that temperature.
- Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- Certain CoolRack modules are SBS-compatible.
Specifica
Specifica
| Capacità | 9 (13 x 75 mm Blood Tubes) |
| Progettazione | CoolRack VS13 |
| Tipo | Blood Collection Tube Module |