Learn More
Copper Nickel wire, 1.6mm (0.064 in.) dia., Thermo Scientific™
Click to view available options
Quantità:
100 m
25 m
5 m
Dimensione della confezione:
100m
25m
5m
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | Copper Nickel wire |
| Nota nome | 1.6mm (0.064 in.) dia. |
| Media Mol. Peso o mole Intervallo di peso | Cu:Ni 55:45 wt% |
| Quantità | 5 m |
| Peso | ≈17.9g/m |
| Forma | Wire |
| Numero MDL | MFCD00801098 |