Learn More
Julabo™ CORIO C-Series Open Heating Bath Circulators with Stainless Steel Baths
BIOPHARMA
Codice prodotto. 15420562
Click to view available options
Modello:
C-B13
C-B17
C-B19
C-B27
C-B5
Capacità (sistema metrico):
13 to 17 L
14 to 19 L
17 to 27 L
3.5 to 5 L
9 to 13 L
Dimensioni (L x P x A):
38 x 23 x 41 cm
40 x 38 x 42 cm
40 x 38 x 47 cm
58 x 38 x 42 cm
58 x 38 x 47 cm
Dimensions (LxWxH) Interior:
15 x 15 x 15 cm (Bath Opening)
18 x 30 x 15 cm (Bath Opening)
18 x 30 x 20 cm (Bath Opening)
35 x 30 x 20 cm (Bath Opening)
35 x 35 x 15 cm (Bath Opening)
Dimensione della confezione:
Pezzo
Codice prodotto. 15420562
Marca: Julabo™ 9011413
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
CORIO C-Series Open Heating Bath Circulators with Stainless Steel Baths are designed for internal applications in the circulator bath. The circulator can be removed for easy cleaning of the bath and a variety of accessories are available.
  • Bright, White, Easy to Read Display
  • Easy to Operate Front Controls
  • Low Liquid Level Early Warning System
  • Integrated Drain Screw (except C-B5)
  • Integrated Timer
  • PID1 and ACC Controls
  • Very Quiet
Specifica
Descrizione Circulation Capacity Flow Rate: 6 L/min., Circulation Capacity Pressure: 0.1 bar
Capacità (sistema metrico) 9 to 13 L
Portata 6 L/min.
Gamma di temperatura 20°C to 100°C (Working)
Tipo Heating Circulator with Open Bath
Da utilizzare con (applicazione) Internal applications at working temperatures from +20 to +100°C
Capacità di riscaldamento 2 kW
Pressione max 0.1 bar
Modello C-B13
Stabilità termica ±0.03°C
Dimensioni (L x P x A) 40 x 38 x 42 cm
Dimensions (LxWxH) Interior 18 x 30 x 15 cm (Bath Opening)