Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Optical Imaging Microplate
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Marca: Corning™ 3985BC
Dettagli aggiuntivi : Peso : 10.79549kg
Descrizione
Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.
Specifica
Standard | |
0.06 cm2 | |
Black, Clear Bottom | |
384 | |
Sterile | |
112 μL | |
With generic barcode | |
20/Pk. | |
Clear |
With Lid | |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
Cell Culture | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |