Learn More
Corning™ BioCoat™ Assay System
Descrizione
Includes
24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifica
Specifica
| Chiusura inclusa | Yes |
| Colore | Clear |
| Area di coltura | 0.3 cm2 |
| Materiale | PET |
| Sterilità | Non-sterile |
| Trattamento superficiale | Collagen |
| Tipo | Cell Culture Insert System |
| Conteggio pozzetti | 24 |
| Da utilizzare con (applicazione) | Rapid differentiation of Caco 2 cells in vitro for robotic screening of prospective pharmaceuticals for oral bioavailability and absorption |
| Include | 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid |
| Vedi altri risultati |