Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
GREENER_CHOICE
Codice prodotto. 10003860
Click to view available options
Diametro (sistema metrico):
35 mm
60 mm
100 mm
150 mm
Quantità:
10/cs.
100/cs.
20/cs.
40/cs.
5/cs.
Dimensione della confezione:
pezzi 10
pezzi 100
pezzi 20
pezzi 40
pezzi 5
Codice prodotto. 10003860
Marca: Corning™ 354468
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
  • Coated with 0.2 micron filtered PDL in a cleanroom
Note: Dishes are not terminally sterilized and not assigned an SAL level.
Order Info
Bulk quantities available; please contact your Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifica
Colore chiusura Clear
Chiusura inclusa Closure Included
Materiale chiusura Polystyrene
Forma del fondo Round
Colore Clear
Area di coltura 21.3 sq. cm
Materiale Polistirene
Forma Round
Trattamento superficiale Poly-D-Lysine
Tipo Cell Culture Dish
Include Lid
Quantità 20/cs.
Diametro (sistema metrico) 60 mm