Learn More
Corning™ CellCube™ Tubing Adaptor
Sfoglia Tutto Corning Prodotti
Click to view available options
Product Type:
4 AseptiQuik™ S Connectors, 4 Tubes Clamps
AseptiQuik™ S Connected to a Female MPC
AseptiQuik™ S Connected to a Male MPC
Dimensione della confezione:
pezzi 4
Descrizione
- Closed System Cell Cube
- CellCube 10, 25, 100 modules, circulation loops, and connectors
- Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifica
Specifica
| Diametro sistema metrico) interno tubo | 9.53 mm |
| Include | Thermoelastic Tubing, Aseptic Connector to Male MPC |
| N. per custodia | 4/Cs. |
| N. per confezione | 1/Pk. |
| Product Type | AseptiQuik™ S Connected to a Male MPC |