Learn More
Corning™ CellCube™ Tubing Manifold
Sfoglia Tutto Corning Prodotti
Click to view available options
:
2/Cs.
Descrizione
- Closed System Cell Cube
- CellCube 10, 25, 100 modules, circulation loops, and connectors
- Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifica
Specifica
| Include | Thermoelastic Tubing, Cross Connection, 4 Aseptic Connectors |
| N. per custodia | 2/Cs. |
| N. per confezione | 1/Pk. |