Learn More
Corning™ Lambda Plus Single-Channel Pipettor, Adjustable, Autoclavable
Sfoglia Tutto Corning Prodotti
Descrizione
Engineered to provide the highest levels of accuracy, reproducibility, and comfort
Available in single-, eight-and twelve-channel configurations, all Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
Features
- Factory calibrated to strict ISO 8655 standards-3-year warranty
- Fully autoclavable and UV resistant
- Designed for universal fit with all common brands of pipet tips
- Adjustable tip ejector
Warranty
3 year
Specifica
Specifica
| Accuracy | ±1.2 to ±0.6% |
| Certificazione/Conformità | ISO8655 |
| Punte compatibili | 200μL Tip |
| Autoclavabile | Autoclavable |
| Da utilizzare con | 200 μL Tip |
| Numero di canali | 1 |
| Product Line | Lambda Plus |
| Tipo | Single-Channel Pipettor |
| Volume (sistema metrico) | Da 20 a 200 μl |
| Da utilizzare con (apparecchiatura) | 200μL Tip |
| Vedi altri risultati |