Learn More
Corning™ Tubing Sets and Connectors for CellCube™
Sfoglia Tutto Corning Prodotti
Click to view available options
:
1/Pk.
Descrizione
- Closed System Cell Cube
- Helps reduce the risk of adventitious contamination
- CellCube 10, 25, 100 modules, circulation loops, and connectors
- Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
Specifica
Specifica
| Include | Medium Filter, 2 Aseptic Connectors |
| N. per custodia | 4/Cs. |
| N. per confezione | 1/Pk. |