Learn More
Corning™ Universal Fit Pipet Tips: Stack Rack, Sterile
Sfoglia Tutto Corning Prodotti
Click to view available options
Volume (sistema metrico):
1 to 200 μL
Dimensione della confezione:
pezzi 4800
Descrizione
- Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
- Beveled orifice helps ensure accurate fluid delivery.
- 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
- Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
- RNase-/DNase-free.
- Nonpyrogenic.
Specifica
Specifica
| Volume (sistema metrico) | 1 to 200 μL |
| Sterilità | Sterile |
| Colore | Natural |
| Formato | In rack |
| Stile punta | Universal |
| Imballaggio | Stack rack |
| Graduato | Yes |
| Materiale | Polipropilene |
| Graduazioni | 10, 50, 100μL |