Learn More
Anti-coronavirus (SARS-CoV-2) spike S2 (B), mouse, mAb, MP Biomedicals™
Monoclonal antibody raised against the coronavirus spike S2 (B) protein, a common target for neutralizing antibodies and vaccines.
Marca: MP Biomedicals 08720412
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Anti-coronavirus (SARS-CoV-2) spike S2 (B), mouse, mAb. The 2019 Novel Coronavirus (SARS-CoV-2) is a coronavirus identified as the cause of an outbreak of respiratory illness first detected in Wuhan, China. Its genome encodes four structural proteins, i.e. spike (S), envelope (E), membrane (M) and nucleocapsid (N). The spike protein (S-protein) contains two subunits, S1 and S2. S1 defines the range of the hosts and specificity of the virus, thus recognizes and binds with the cell surface receptor. S2 subunit contains basic elements needed for the membrane fusion. The spike protein is the common target for neutralizing antibodies and vaccines.
Specifica
| Coronavirus Spike S2 (B) Protein | |
| Monoclonal | |
| Mouse | |
| RUO | |
| None |
| Western Blot | |
| Unconjugated | |
| 250 μg | |
| Primary | |
| IgG1 |
Titolo del prodotto
For research or further manufacturing use only, not for direct human use.
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto