Learn More
Hirschmann™ Camere di conteggio
Click to view available options
Product Type:
Counting Chambers, Bright lined with clamps; Neubauer-improved
Counting Chambers, Bright lined; Neubauer-improved
Counting Chambers, Neubauer
Counting Chambers, Neubauer-improved
Dimensione della confezione:
Pezzo
Descrizione
- Double net ruling
- Available bright-lined or no, with or without clamps
- Includes 2 cover glasses for haemocytometer
- CE acc. IVD 98/79 EC
Specifica
Specifica
| Product Type | Counting Chambers, Bright lined with clamps; Neubauer-improved |
| Quantità | 1Each |
| Profondità (sistema metrico) | 0.1mm |
| Include | 2 Haemocytometer Glass Covers and Clamps |