Learn More
Hirschmann™ Counting Chambers
Click to view available options
Product Type:
Camere di conteggio
Counting Chamber (Burker)
Counting Chamber (Fuch-Rosenthal)
Counting Chamber (Nageotte)
Counting Chamber (Neubauer)
Counting Chamber (Thoma)
Counting Chamber (Thoma-neu)
Descrizione:
Double Net Ruling, Bright Lined
Double Net Ruling, Clamps
Double Net Ruling, Clamps, Bright Lined
Dimensione della confezione:
Pezzo
Descrizione
- Double net ruling
- Including 2 cover glasses for hemacytometer
- CE acc. IVD 98/79 EC
Specifica
Specifica
| Product Type | Counting Chamber (Fuch-Rosenthal) |
| Descrizione | Double Net Ruling, Clamps |
| Quantità | 1 Each |
| Profondità (sistema metrico) | 0.2 mm |
| Certificazione/Conformità | CE acc. IVD 98/79 EC |
| Include | 2 cover glasses for haemacytometer |