Learn More
Hirschmann™ Counting Chambers with Clamps
Click to view available options
Product Type:
Counting Chamber (Burker)
Counting Chamber (Neubauer)
Dimensione della confezione:
Pezzo
Descrizione
- Double net ruling
- With clamps
- Includes 2 haemocytometer glass covers
- With LOT number on packaging
- Labelled with: usable until, CE pursuant to IVD 98/79 EC
Specifica
Specifica
| Product Type | Counting Chamber (Neubauer) |
| Descrizione | Double net ruling |
| Profondità (sistema metrico) | 0.1 mm |
| Certificazione/Conformità | CE pursuant to IVD 98/79 EC |
| Include | 2 haemocytometer glass covers |