Learn More
Sonics Materials Accoppiatore
Click to view available options
Diametro (sistema metrico):
2 mm
Dimensione della confezione:
Pezzo
Descrizione
- Stepped microtip/probe assembly
- Coupler - 33/4 in. (95 mm)
- The stepped microtip/probe assembly which consists of two parts, the coupler (standard or reverse) and the microtip or probe, screws into the converter in place of the probe
- Capable of reaching into narrower vessels than the tapered microtip, the stepped microtip assembly can process volumes as small as 150 μl
- Microtips are fabricated from titanium alloy Ti-6Al-4V and are autoclavable
Specifica
Specifica
| Product Type | Accoppiatore |
| Diametro (sistema metrico) | 2 mm |
| Lunghezza (sistema metrico) | 9.5 mm |
| Quantità confezione | 1 ciascuno |