Learn More
Euro Protection™ Coverguard™ Okenite Clog
Click to view available options
:
Color: White; Material: Microfiber
Descrizione
Order Info
dimColor
Specifica
Specifica
| Certificazione/Conformità | CE Certified, EN ISO 20345 |
| Colore | White |
| Materiale pianta del piede | Double density Polyurethane |
| Materiale | Microfiber |
| Size | 37 |
| Caratteristiche suola | Double density Polyurethane construction |
| Materiale suola | Double density Polyurethane |
| Materiale dello zoccolo | Steel |
| Resistente a | Oil and hydrocarbon |
| Product Type | Clog |