Learn More
COX7C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11411-2-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody is specific to COX7C.
Cytochrome c oxidase (COX), the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may function in the regulation and assembly of the complex.Specifica
| COX7C | |
| Polyclonal | |
| Unconjugated | |
| COX7C | |
| COX7C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100188937, 12867, 1350 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P15954, P17665, P80432 | |
| COX7C | |
| COX7C Fusion Protein Ag1989 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |