Learn More
Cole-Parmer™ CPC™ DrumQuik PRO System Pro Adapter
Click to view available options
Dimensioni dei tubi:
1/2 in. NPTF x 1/2 in. Hose Barb
1/2 in. NPTF x 1/2 in. OD
Dimensione della confezione:
Pezzo
Descrizione
- Easy-to-use, economical solution for chemical dispensing.
- Durable materials withstand harsh operating conditions.
- Ideal for 5-gallon jerry cans, 55-gallon drums and IBCs.
- The DrumQuik PRO system offers a simplified solution to your chemical dispensing needs.
- Maintains a closed system, protecting your facility, your operating environment, and your employees from the potential dangers of chemical drips, spills and escaping fumes.
- Simple connections, rugged construction, and chemical-resistant materials handle a wide variety of applications including acids and caustics, agricultural liquids, cleaning agents and sterilants, paints and coatings and food oils and syrups.
- Base components are a reusable quick-disconnect coupler and a single-use drum insert assembly; both of which are made from environmentally friendly, food-grade polyethylene (PE).
- Order a convenient dispensing kit which includes all required components, or customize your own system with individual components.
- Coupler body and plugs mate equally well with all drum inserts listed.
Specifica
Specifica
| Descrizione | Barb x NPT(F) |
| Materiale | Polypropylene |
| Dimensioni dei tubi | 1/2 in. NPTF x 1/2 in. Hose Barb |
| Portata | 10 GPM at less than 2 psid (0.14 bar) pressure drop |
| Pressione massima | 45 psi |
| Product Type | Pro Adapter, 1/2 in. NPTF x 1/2 in. Hose Barb |