Learn More
Cole-Parmer™ CPC™ NS4 Non-spill Panel Mount Coupling Insert with Compression Connection
Click to view available options
Diametro (sistema imperiale) esterno:
3/8 in.
Dimensione della confezione:
Pezzo
Descrizione
- Less than 0.10mL spillage.
- One-hand operation.
- Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance.
- For a complete fitting, order one body and one insert.
- All bodies will mate with inserts to make a fitting that will work for multiple applications.
Specifica
Specifica
| Tipo | Coupling Insert, Non-Spill, Panel Mount, 3/8 in. OD Tube |
| Diametro (sistema imperiale) esterno | 3/8 in. |
| Descrizione | Quick-Disconnect x Compression |
| Materiale | ABS Plastic |