Learn More
Cole-Parmer™ CPC™ NSH Non-spill Coupling Insert with NPT(M) Thread
Codice prodotto. 15417373
Click to view available options
Dimensioni dei tubi:
1/2 in.
Dimensione della confezione:
Pezzo
Codice prodotto. 15417373
Marca: Cole-Parmer™ 3160372
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
  • NS6 Series couplings feature non-spill valves at a great price.
  • Use the NS6 when even a few drops of spillage pose problems regarding safety, media cost or environmental regulations.
  • Soft touch overmold makes these couplings comfortable in the hand and very attractive.
  • Lightweight, chemically resistant and easy to use.
  • Non-spill design.
  • Disconnect under pressure with no spills.
  • Color coding Instant visual differentiation of media lines.
  • Glass-filled polypropylene.
  • Durable and compatible with many chemicals.
  • Colder thumb latch.
  • One-hand connection and disconnection.
Specifica
Descrizione Quick-Disconnect x NPTM(M)
Materiale Polypropylene
Dimensioni dei tubi 1/2 in.
Flessibile flangiato 1/2 in.
Porta di uscita 1/2 in. Quick Disconnect
Product Type Non-Spill, Threaded, Quick-Disconnect x NPT(M) Coupling Insert