Learn More
CPP Binding Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca:  Invitrogen PA3107
Codice prodotto. 11518761
  • 508.00€ / 200µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
PA3-107 detects the CPP-binding protein. PA3-107 has been used successfully in Western blot, ELISA, immunoprecipitation, immunohistochemistry, and immunocytochemistry procedures. In Western blot analysis of rat brain synaptic membranes this antibody detects ∽80 kDa and ∽60 kDa proteins representing CPP-binding protein. The ∽60kDa protein is likely a breakdown product of the ∽80 kDa CPP-binding protein. The PA3-107 immunogen is a synthetic peptide corresponding to the C-terminal residues K(396) A E P K R T T L G(405) C of rat CPP-binding protein.
CPP is a potent antagonist acting selectively at NMDA type excitatory amino acid receptors. This compound is more potent than previously reported NMDA antagonists in depressing mammalian spinal neuronal responses.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
CPP Binding Protein
Polyclonal
Rabbit
200 μL
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Synthetic peptide corresponding to the C-terminal residues K(396) A E P K R T T L G(405) C of rat CPP-binding protein.
RUO
Human, Mouse, Rat