Learn More
CPP Binding Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA3107
Descrizione
PA3-107 detects the CPP-binding protein. PA3-107 has been used successfully in Western blot, ELISA, immunoprecipitation, immunohistochemistry, and immunocytochemistry procedures. In Western blot analysis of rat brain synaptic membranes this antibody detects ∽80 kDa and ∽60 kDa proteins representing CPP-binding protein. The ∽60kDa protein is likely a breakdown product of the ∽80 kDa CPP-binding protein. The PA3-107 immunogen is a synthetic peptide corresponding to the C-terminal residues K(396) A E P K R T T L G(405) C of rat CPP-binding protein.
CPP is a potent antagonist acting selectively at NMDA type excitatory amino acid receptors. This compound is more potent than previously reported NMDA antagonists in depressing mammalian spinal neuronal responses.
Specifica
| CPP Binding Protein | |
| Polyclonal | |
| Rabbit | |
| 200 μL | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Synthetic peptide corresponding to the C-terminal residues K(396) A E P K R T T L G(405) C of rat CPP-binding protein. | |
| RUO | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |