missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase, Muscle/CKMM Mouse anti-Human, Clone: CKMM/3341, Novus Biologicals™
Mouse Monoclonal Antibody
299.00€
Specifica
| Antigene | Creatine Kinase, Muscle/CKMM |
|---|---|
| Clone | CKMM/3341 |
| Concentrazione | 0.2 mg/ml |
| Diluizione | ELISA, Protein Array |
| Applicazioni | ELISA, Peptide Array |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18394313
|
Novus Biologicals
NBP3-07560-20UG |
20 μg |
299.00€
20µg |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Description
Creatine Kinase, Muscle/CKMM Monoclonal specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for ELISA, Protein Array.Specifications
| Creatine Kinase, Muscle/CKMM | |
| 0.2 mg/ml | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
| Recombinant full-length Creatine Kinase, Muscle/CKMM protein | |
| Primary | |
| Store at 4°C. |
| CKMM/3341 | |
| ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1158 | |
| IgG2a κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title