Learn More
Thermo Scientific Chemicals Cresol Red sodium salt
Descrizione
Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cresol Red sodium salt acts as a constituent of polymerase chain reaction reagent systems. It is useful as a pH indicator with color changes with pH 0.2 - pH 1.8 and pH 7.1 - 8.8. Further, it plays an important role in molecular biology reactions in place of other loading dyes.
Solubility
Soluble in water, dilute alcohols, acids, alkalies and ethanol.
Notes
Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red)Incompatible with strong oxidizing agents.
Specifica
Specifica
| Materiale o nome chimico | Cresol Red sodium salt |
| Punto di fusione | ∼250°C (decomposition) |
| Formula molecolare | C21H17NaO5S |
| Informazioni di solubilità | Soluble in water,dilute alcohols,acids,alkalies and ethanol. |
| Peso formulazione | 404.42 |
| Quantità | 25 g |
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.