Learn More
Skytec Crest Grip Gloves
Click to view available options
Size:
7, Small
8, Medium
9, Large
10, X-Large
11, 2X-Large
Dimensione della confezione:
10 paia
Descrizione
- GeniumTM is an eco friendly liner with 1/3 of the ingredients by weight that are sustainably sourced and that requires 30% less energy to be produced
- Crinkle effect on a nitrile foam providing oustanding grip in all conditions
- Stretch with recovery around 2-3x greater than nylon
- Superior abrasion resistance up to 18,000 Cycles
- No Particle shedding
- Super soft liner
- Anatomically designed with moisture wicking to deliver great comfort
- Sanitized™ antimicrobial finish provides extra freshness
- Light heat protection up to 100°C for a short period
Specifica
Specifica
| ANSI Abrasion Rating | ANSI 3 |
| Certificazione/Conformità | CE CAT II |
| Certifications/Compliance (Europe) | EN388, EN420, EN 407 |
| Chlorinated | No |
| Area rivestita | Palm |
| Struttura | Knit |
| Coating Material | Nitrile Foam |
| Colore | Red, Black |
| Finitura esterna | Partially Textured |
| Hand | Specific |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido